Assay Details
Target Gene Details
Entrez Gene ID: | 258601 |
Gene Name: | olfactory receptor 984 |
Gene Aliases: |
MOR239-6 |
Location: |
Chr.9:40100544-40156029 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr984 | NM_146608.1 | 1 | 163 | NP_666819.1 |
More Information
Set Membership: |
|

Genomic Map