Assay Details
Target Gene Details
Entrez Gene ID: | 258601 |
Gene Name: | olfactory receptor 984 |
Gene Aliases: |
MOR239-6 |
Location: |
Chr.9:40100544-40156029 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr984 |
Target Gene Details
Entrez Gene ID: | 258854 |
Gene Name: | olfactory receptor 985 |
Gene Aliases: |
MOR171-4 |
Location: |
Chr.9:40126924-40128039 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Olfr985 | NM_146855.2 | 1 | 709 | NP_667066.1 |
| BC138430.1 | 1 | 709 | ||
| BC138431.1 | 1 | 709 |
More Information
Set Membership: |
|

Genomic Map