Assay Details
Target Gene Details
Entrez Gene ID: | 20587 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1 |
Gene Aliases: |
AU020204, Baf47, Ini1, SNF5/INI1, Snf5 |
Location: |
Chr.10:75896769-75921614 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smarcb1 | NM_001161853.1 | 9 | 1423 | NP_001155325.1 |
| NM_011418.2 | 9 | 1450 | NP_035548.1 | |
| AB041578.1 | 9 | 1406 | BAA95062.1 | |
| AI840539.1 | 1 | 225 | ||
| AJ011739.1 | 9 | 1424 | CAA09760.1 | |
| AJ011740.1 | 9 | 1424 | CAA09761.1 | |
| AK008558.1 | 7 | 1155 | ||
| AK075906.1 | 9 | 1433 | BAC36045.1 | |
| AK145552.1 | 9 | 1412 | BAE26504.1 | |
| AK147638.1 | 5 | 7239 | ||
| AK147685.1 | 5 | 7239 | ||
| AK150169.1 | BAE29357.1 | |||
| AK205590.1 | 2 | 136 | ||
| AK208680.1 | 2 | 136 | ||
| BC025163.1 | 9 | 1404 | AAH25163.1 |
More Information
Set Membership: |
|

Genomic Map