Assay Details
Target Gene Details
Entrez Gene ID: | 11705 |
Gene Name: | anti-Mullerian hormone |
Gene Aliases: |
MIS |
Location: |
Chr.10:80805021-80807679 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Amh | NM_007445.2 | 5 | 1382 | NP_031471.2 |
| AY911505.1 | 5 | 1382 | AAW84293.1 | |
| BC150477.1 | 5 | 1381 |

Genomic Map