Assay Details
Target Gene Details
Entrez Gene ID: | 18119 |
Gene Name: | nodal |
Gene Aliases: |
Tg.413d |
Location: |
Chr.10:61417972-61425337 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nodal | NM_013611.4 | 1 | 261 | NP_038639.2 |
| X70514.1 | 1 | 261 | CAA49914.1 |
More Information
Set Membership: |
|

Genomic Map