Assay Details
Target Gene Details
Entrez Gene ID: | 212862 |
Gene Name: | choline phosphotransferase 1 |
Gene Aliases: |
- |
Location: |
Chr.10:88472813-88503970 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chpt1 | NM_001146690.1 | 1 | 200 | NP_001140162.1 |
| NM_144807.3 | 1 | 200 | NP_659056.3 | |
| NR_027477.1 | 1 | 200 | ||
| AK007964.1 | 1 | 121 | ||
| AK032497.1 | 1 | 95 | BAC27897.1 | |
| AY445814.1 | 1 | 95 | AAR16089.1 | |
| BC016251.1 | 1 | 148 | AAH16251.1 | |
| BC031435.1 | 1 | 155 | AAH31435.1 | |
| BY341208.1 | 1 | 108 | ||
| CJ177670.1 | 1 | 200 |
More Information
Set Membership: |
|

Genomic Map