Assay Details
Target Gene Details
Entrez Gene ID: | 23943 |
Gene Name: | extended synaptotagmin-like protein 1 |
Gene Aliases: |
Fam62a, Mbc2, vp115 |
Location: |
Chr.10:128510248-128525859 on Build GRCm38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Esyt1 | NM_011843.2 | 31 | 3485 | NP_035973.1 |
| AF098633.1 | 8 | 962 | AAD10189.1 | |
| AF098634.1 | 22 | 2278 | AAD10190.1 | |
| AK008224.1 | 5 | 587 | BAB25542.1 | |
| AK054147.1 | 21 | 2361 | ||
| AK088421.1 | 31 | 3500 | ||
| AK189859.1 | ||||
| AK198125.1 | ||||
| BC011482.1 | 31 | 3485 | AAH11482.1 |

Genomic Map