Assay Details
Target Gene Details
Entrez Gene ID: | 71865 |
Gene Name: | F-box protein 30 |
Gene Aliases: |
1700026A16Rik, MUSA1 |
Location: |
Chr.10:11281330-11298048 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fbxo30 | NM_001168297.1 | 2 | 1852 | NP_001161769.1 |
| NM_027968.3 | 2 | 1886 | NP_082244.2 | |
| AK006369.1 | 2 | 1853 | BAB24551.1 | |
| AK083479.1 | 2 | 1843 | BAC38931.1 | |
| AK135274.1 | 2 | 1845 | BAE22469.1 | |
| BC037691.1 | 1 | 765 | AAH37691.1 |

Genomic Map