Assay Details
Target Gene Details
Entrez Gene ID: | 100504069 |
Gene Name: | predicted gene 15915 |
Gene Aliases: |
OTTMUSG00000027787 |
Location: |
Chr.10:93674218-93683322 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm15915 | NR_038017.1 | |||
| AK131848.1 |
Target Gene Details
Entrez Gene ID: | 57764 |
Gene Name: | netrin 4 |
Gene Aliases: |
- |
Location: |
Chr.10:93641049-93745972 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ntn4 | NM_021320.3 | 3 | 1037 | NP_067295.2 |
| AF268066.1 | 3 | 632 | AAF91404.1 | |
| AF281278.1 | 3 | 885 | AAG30823.1 | |
| BC119182.1 | 3 | 719 | ||
| BC119184.1 | 3 | 719 |
More Information
Set Membership: |
|

Genomic Map