Assay Details
Target Gene Details
Entrez Gene ID: | 238276 |
Gene Name: | A kinase (PRKA) anchor protein 5 |
Gene Aliases: |
3526401B18Rik, AKAP 150, AKAP-5, AKAP150, BB098886, Gm258, P150 |
Location: |
Chr.12:76324402-76334151 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akap5 | NM_001101471.1 | 2 | 2289 | NP_001094941.1 |
More Information
Set Membership: |
|

Genomic Map