Assay Details
Target Gene Details
Entrez Gene ID: | 14025 |
Gene Name: | B cell CLL/lymphoma 11A (zinc finger protein) |
Gene Aliases: |
2810047E18Rik, BCL-11A, Ctip1, D930021L15Rik, Evi9, Evi9a, Evi9b, Evi9c, mKIAA1809 |
Location: |
Chr.11:24076564-24173558 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bcl11a | NM_001159289.1 | 5 | 1387 | NP_001152761.1 |
| NM_001159290.1 | 5 | 1042 | NP_001152762.1 | |
| NM_016707.3 | 5 | 2987 | NP_057916.1 | |
| AF051525.1 | AAF22430.1 | |||
| AF169036.1 | AAF65928.1 | |||
| AF169037.1 | AAF65929.1 | |||
| AK004395.1 | BAB23285.1 | |||
| AK043677.1 | 5 | 1186 | BAC31616.1 | |
| AK045556.1 | 5 | 1043 | BAC32416.1 | |
| BC010585.1 | AAH10585.1 | |||
| BC051418.1 | AAH51418.1 |
More Information
Set Membership: |
|

Genomic Map