Assay Details
Target Gene Details
Entrez Gene ID: | 74476 |
Gene Name: | RIKEN cDNA 4933439C10 gene |
Gene Aliases: |
2610028M21Rik |
Location: |
Chr.11:59505685-59511067 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4933439C10Rik | NR_015585.2 | 1 | 876 | |
| AK011602.1 | 1 | 409 | ||
| AK017115.1 | 1 | 877 | ||
| AK163429.1 | 1 | 866 | ||
| AK169756.1 | 1 | 909 | BAE41348.1 |
Target Gene Details
Entrez Gene ID: | 268417 |
Gene Name: | zinc finger with KRAB and SCAN domains 17 |
Gene Aliases: |
Nizp1, Zfp496, Znf496 |
Location: |
Chr.11:59485520-59506640 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zkscan17 | NM_001130529.2 | 1 | 81 | NP_001124001.1 |
| NM_001291014.1 | 1 | 81 | NP_001277943.1 | |
| NM_172941.4 | 1 | 81 | NP_766529.3 | |
| AK139346.1 | 1 | 321 | BAE23969.1 | |
| CB528232.1 | 1 | 87 |
More Information
Set Membership: |
|

Genomic Map