Assay Details
Target Gene Details
Entrez Gene ID: | 11782 |
Gene Name: | adaptor-related protein complex AP-4, sigma 1 |
Gene Aliases: |
AI314282 |
Location: |
Chr.12:51689888-51738939 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap4s1 | NM_021710.3 | 6 | 850 | NP_068356.1 |
| AF092093.1 | 6 | 779 | AAD20447.1 | |
| AK005283.1 | 6 | 600 | BAB23931.1 | |
| AK178071.1 | ||||
| BC001985.1 | AAH01985.1 | |||
| BC053339.1 | 6 | 610 | AAH53339.1 | |
| BP426557.1 | 4 | 305 |
More Information
Set Membership: |
|

Genomic Map