Assay Details
Target Gene Details
Entrez Gene ID: | 14457 |
Gene Name: | growth arrest specific 7 |
Gene Aliases: |
AW124766, B230343A10Rik, Gas7-cb |
Location: |
Chr.11:67455024-67688992 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gas7 | NM_001109657.3 | 14 | 4271 | NP_001103127.1 |
| NM_001277079.1 | 14 | 4100 | NP_001264008.1 | |
| NM_001277080.1 | 13 | 4014 | NP_001264009.1 | |
| NM_008088.5 | 14 | 4077 | NP_032114.2 | |
| AK152149.1 | BAE30985.1 |
Target Gene Details
Entrez Gene ID: | 93896 |
Gene Name: | glucagon-like peptide 2 receptor |
Gene Aliases: |
9530092J08Rik, GLP-2 |
Location: |
Chr.11:67663299-67771153 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glp2r |
More Information
Set Membership: |
|

Genomic Map