Assay Details
Target Gene Details
Entrez Gene ID: | 66569 |
Gene Name: | glycerophosphodiester phosphodiesterase domain containing 1 |
Gene Aliases: |
2610020H15Rik |
Location: |
Chr.11:87033794-87074953 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gdpd1 | NM_025638.2 | 1 | 251 | NP_079914.1 |
| AK011487.1 | 1 | 176 | BAB27650.1 | |
| AK013864.1 | 1 | 177 | BAB29022.1 | |
| AK016023.1 | 1 | 177 | BAB30083.1 | |
| AK167812.1 | 1 | 234 | BAE39839.1 | |
| BC016541.1 | 1 | 174 | AAH16541.1 | |
| BY282485.1 | 1 | 252 |
More Information
Set Membership: |
|

Genomic Map