Assay Details
Target Gene Details
Entrez Gene ID: | 216578 |
Gene Name: | poly(A) polymerase gamma |
Gene Aliases: |
9630006B20Rik |
Location: |
Chr.11:23860145-23895296 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Papolg | NM_172555.2 | 22 | 3528 | NP_766143.2 |
| AK035810.1 | 22 | 3511 | BAC29194.1 | |
| BC019635.1 | 7 | 1880 | AAH19635.1 | |
| BC023030.1 | 7 | 1865 | AAH23030.1 | |
| BC059271.1 | 22 | 3522 | AAH59271.1 |
More Information
Set Membership: |
|

Genomic Map