Assay Details
Target Gene Details
Entrez Gene ID: | 69747 |
Gene Name: | zinc finger SWIM-type containing 7 |
Gene Aliases: |
2410012H22Rik |
Location: |
Chr.11:62267224-62281395 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zswim7 | NM_027198.1 | 5 | 436 | NP_081474.1 |
| AA027411.1 | ||||
| AK010472.1 | 5 | 416 | BAB26965.1 | |
| BC109343.2 | 5 | 394 | AAI09344.1 | |
| BQ554850.1 | 5 | 439 |

Genomic Map