Assay Details
Target Gene Details
Entrez Gene ID: | 237911 |
Gene Name: | BRCA1 interacting protein C-terminal helicase 1 |
Gene Aliases: |
3110009N10Rik, 8030460J03Rik, Bach1, FACJ, Fancj, OF |
Location: |
Chr.11:86058131-86201267 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Brip1 | NM_178309.2 | 7 | 854 | NP_840094.1 |
| AK033205.1 | 7 | 826 | ||
| AK051878.1 | 7 | 855 | BAC34798.2 | |
| AK145709.1 | 7 | 855 | BAE26603.1 | |
| AK169440.1 | 7 | 801 | BAE41179.1 | |
| AK169823.1 | 7 | 813 | BAE41393.1 | |
| BC094252.1 | 7 | 803 | AAH94252.1 |
More Information
Set Membership: |
|

Genomic Map