Assay Details
Target Gene Details
Entrez Gene ID: | 15184 |
Gene Name: | histone deacetylase 5 |
Gene Aliases: |
AI426555, Hdac4, mHDA1, mKIAA0600 |
Location: |
Chr.11:102195747-102230186 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hdac5 | NM_001077696.1 | 4 | 678 | NP_001071164.1 |
| NM_001284248.1 | 6 | 1190 | NP_001271177.1 | |
| NM_001284249.1 | 5 | 787 | NP_001271178.1 | |
| NM_001284250.1 | 4 | 675 | NP_001271179.1 | |
| NM_010412.3 | 4 | 675 | NP_034542.3 | |
| AF006602.2 | 3 | 391 | AAD09834.2 | |
| AF207748.1 | 3 | 310 | AAF31418.1 | |
| AK139202.1 | 5 | 756 | ||
| AK145972.1 | 4 | 640 | ||
| AK146483.1 | 4 | 614 | BAE27204.1 | |
| AK155335.1 | 4 | 649 | BAE33201.1 | |
| AK156008.1 | 2 | 728 | ||
| AK157159.1 | 2 | 728 | ||
| AK194061.1 | ||||
| AK194600.1 | ||||
| BC060609.1 | 3 | 469 | AAH60609.1 |
More Information
Set Membership: |
|

Genomic Map