Assay Details
Target Gene Details
Entrez Gene ID: | 16665 |
Gene Name: | keratin 15 |
Gene Aliases: |
AI528832, K15, Krt1-15 |
Location: |
Chr.11:100131759-100135949 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt15 | NM_008469.2 | 1 | 94 | NP_032495.2 |
| BC057934.1 | 1 | 73 | AAH57934.1 | |
| BY061070.1 | 1 | 95 | ||
| BY107287.1 | 1 | 95 |

Genomic Map