Assay Details
Target Gene Details
Entrez Gene ID: | 22361 |
Gene Name: | vanin 1 |
Gene Aliases: |
V-1 |
Location: |
Chr.10:23894688-23905344 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vnn1 | NM_011704.3 | 1 | 342 | NP_035834.2 |
| AJ132098.1 | 1 | 175 | CAA10567.1 | |
| AK002773.1 | 1 | 174 | BAB22347.1 | |
| AK145984.1 | 1 | 248 | BAE26806.1 | |
| AK167427.1 | 1 | 174 | BAE39515.1 | |
| BB838710.1 | 1 | 342 | ||
| BC019203.1 | 1 | 168 | AAH19203.1 |
More Information
Set Membership: |
|

Genomic Map