Assay Details
Target Gene Details
Entrez Gene ID: | 93736 |
Gene Name: | AF4/FMR2 family, member 4 |
Gene Aliases: |
AF5Q31, Alf4, Laf4l, MCEF |
Location: |
Chr.11:53350307-53421830 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aff4 | NM_033565.2 | 11 | 1899 | NP_291043.1 |
| AF190449.1 | 11 | 1841 | AAG17126.1 | |
| AK044058.1 | 11 | 1783 | ||
| AK054401.1 | 11 | 1747 | BAC35763.1 | |
| AK163213.1 | 1 | 357 | ||
| BC138999.1 | 11 | 1572 | ||
| BC145490.1 | 10 | 1485 |
More Information
Set Membership: |
|

Genomic Map