Assay Details
Target Gene Details
Entrez Gene ID: | 12799 |
Gene Name: | 2',3'-cyclic nucleotide 3' phosphodiesterase |
Gene Aliases: |
CNPase, Cnp-1, Cnp1 |
Location: |
Chr.11:100574909-100591521 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnp | NM_001146318.1 | 1 | 682 | NP_001139790.1 |
| NM_009923.2 | 2 | 467 | NP_034053.2 | |
| AF332055.1 | 1 | 280 | AAK56084.1 | |
| AF332056.1 | 1 | 280 | AAK56085.1 | |
| AK041969.1 | 1 | 683 | ||
| AK050628.1 | 2 | 432 | BAC34351.1 | |
| AK087415.1 | 2 | 433 | ||
| AK140280.1 | 1 | 683 | BAE24314.1 | |
| AK154654.1 | 2 | 491 | BAE32745.1 | |
| AK158318.1 | 2 | 461 | BAE34457.1 | |
| AK158520.1 | 2 | 382 | BAE34543.1 | |
| AK172318.1 | 2 | 432 | ||
| AK172514.1 | 2 | 432 | ||
| BC005544.1 | 2 | 420 | AAH05544.1 | |
| BC021904.1 | 2 | 356 | AAH21904.1 | |
| M31810.1 | 1 | 1049 | AAA37429.1 | |
| M58045.1 | 2 | 390 | AAA37430.1 |
More Information
Set Membership: |
|

Genomic Map