Assay Details
Target Gene Details
Entrez Gene ID: | 12799 |
Gene Name: | 2',3'-cyclic nucleotide 3' phosphodiesterase |
Gene Aliases: |
CNPase, Cnp-1, Cnp1 |
Location: |
Chr.11:100574909-100591521 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cnp | NM_001146318.1 | 3 | 1619 | NP_001139790.1 |
| NM_009923.2 | 4 | 1404 | NP_034053.2 | |
| AF332055.1 | AAK56084.1 | |||
| AF332056.1 | AAK56085.1 | |||
| AK041969.1 | 3 | 1619 | ||
| AK050628.1 | BAC34351.1 | |||
| AK087415.1 | 4 | 1370 | ||
| AK140280.1 | 3 | 1620 | BAE24314.1 | |
| AK154654.1 | 4 | 1428 | BAE32745.1 | |
| AK158318.1 | 4 | 1398 | BAE34457.1 | |
| AK158520.1 | 4 | 1319 | BAE34543.1 | |
| AK172318.1 | 4 | 1369 | ||
| AK172514.1 | 4 | 1369 | ||
| BC005544.1 | 4 | 1357 | AAH05544.1 | |
| BC021904.1 | 4 | 1293 | AAH21904.1 | |
| M31810.1 | 3 | 1985 | AAA37429.1 | |
| M58045.1 | 4 | 1327 | AAA37430.1 |
More Information
Set Membership: |
|

Genomic Map