Assay Details
Target Gene Details
Entrez Gene ID: | 66201 |
Gene Name: | Vps20-associated 1 homolog (S. cerevisiae) |
Gene Aliases: |
1110001D18Rik, 1110059P08Rik, AU040813, C85340, LIP5, SBP1 |
Location: |
Chr.10:14654760-14705527 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vta1 | NM_025418.3 | 8 | 896 | NP_079694.2 |
| AK002611.1 | 8 | 895 | BAB22228.1 | |
| AK003213.1 | 7 | 813 | ||
| AK004323.1 | 8 | 968 | BAE43171.1 | |
| AK009227.2 | 8 | 885 | BAB26150.1 | |
| AK042917.1 | 2 | 131 | ||
| BC026752.1 | 8 | 889 | AAH26752.1 |
More Information
Set Membership: |
|

Genomic Map