Assay Details
Target Gene Details
Entrez Gene ID: | 19739 |
Gene Name: | regulator of G-protein signaling 9 |
Gene Aliases: |
RGS9-1, Rgs9-2 |
Location: |
Chr.11:109225348-109298181 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Rgs9 | NM_011268.2 | 19 | 2204 | NP_035398.2 |
AF125046.1 | 19 | 2160 | AAD20014.1 | |
BC129898.1 | 11 | 1482 | ||
BC129899.1 | 19 | 2070 |
