Assay Details
Target Gene Details
Entrez Gene ID: | 55963 |
Gene Name: | solute carrier family 1 (glutamate/neutral amino acid transporter), member 4 |
Gene Aliases: |
ASCT-1, ASCT1, AW045657, SATT |
Location: |
Chr.11:20302180-20332713 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc1a4 | NM_018861.3 | 8 | 3156 | NP_061349.3 |
| AK136900.1 | 8 | 3156 | BAE23162.1 | |
| AK140866.1 | 8 | 3157 | BAE24502.1 | |
| AK148075.1 | 8 | 3157 | BAE28327.1 | |
| AK190214.1 | 1 | 332 | ||
| BC043483.1 | 8 | 3114 | AAH43483.1 | |
| BC052733.1 | 8 | 3030 | AAH52733.1 | |
| U75215.1 | 8 | 3026 | AAB71740.1 |
More Information
Set Membership: |
|

Genomic Map