Assay Details
Target Gene Details
Entrez Gene ID: | 212706 |
Gene Name: | NEDD4 binding protein 3 |
Gene Aliases: |
C330016O10Rik |
Location: |
Chr.11:51643071-51651073 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| N4bp3 | NM_145974.3 | 3 | 833 | NP_666086.2 |
| AK049249.1 | 3 | 603 | BAC33636.1 | |
| AK077848.1 | 3 | 596 | ||
| AK141228.1 | 3 | 627 | BAE24601.1 | |
| AK142500.1 | 3 | 596 | BAE25088.1 | |
| AK170270.1 | 3 | 631 | BAE41675.1 | |
| AK170521.1 | 3 | 628 | BAE41855.1 | |
| BC006740.1 | 3 | 546 | AAH06740.2 | |
| BC055831.1 | 3 | 833 | AAH55831.2 |

Genomic Map