Assay Details
Target Gene Details
Entrez Gene ID: | 18102 |
Gene Name: | NME/NM23 nucleoside diphosphate kinase 1 |
Gene Aliases: |
AL024257, NDPK-A, NM23-M1, NM23A |
Location: |
Chr.11:93958925-93968521 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nme1 | NM_008704.2 | 1 | 100 | NP_032730.1 |
| BY037001.1 | 1 | 100 |
More Information
Set Membership: |
|

Genomic Map