Assay Details
Target Gene Details
Entrez Gene ID: | 72129 |
Gene Name: | peroxisomal biogenesis factor 13 |
Gene Aliases: |
2610008O20Rik |
Location: |
Chr.11:23646479-23665935 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pex13 | NM_023651.4 | 4 | 2565 | NP_076140.2 |
| AK029336.1 | 4 | 2538 | BAC26402.1 | |
| AK031710.1 | 4 | 2565 | BAC27526.1 | |
| AK031975.1 | 4 | 2521 | BAC27635.1 | |
| AK032650.1 | 3 | 1934 | BAC27971.1 |
More Information
Set Membership: |
|

Genomic Map