Assay Details
Target Gene Details
Entrez Gene ID: | 216984 |
Gene Name: | ecotropic viral integration site 2b |
Gene Aliases: |
Evi-2, Evi2 |
Location: |
Chr.11:79513385-79523762 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Evi2b | NM_001077496.1 | 2 | 1279 | NP_001070964.1 |
| AK039968.1 | 2 | 1256 | BAE20560.1 | |
| AK170476.1 | 2 | 1241 | BAE41820.1 | |
| AK171686.1 | 2 | 1196 | BAE42611.1 | |
| BC017548.1 | 2 | 1228 | AAH17548.1 |
Target Gene Details
Entrez Gene ID: | 101488212 |
Gene Name: | predicted gene 21975 |
Gene Aliases: |
Evi2a-evi2b |
Location: |
Chr.11:79513385-79530609 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm21975 | NM_146023.4 | 4 | 1714 | NP_666135.1 |
Target Gene Details
Entrez Gene ID: | 18015 |
Gene Name: | neurofibromatosis 1 |
Gene Aliases: |
AW494271, E030030H24Rik, Nf-1 |
Location: |
Chr.11:79335988-79581609 on Build GRCm38 |
Assay Gene Location: | Within Intron 36 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nf1 | NM_010897.2 | NP_035027.1 | ||
| L10370.1 | AAA68132.1 |
More Information
Set Membership: |
|

Genomic Map