Assay Details
Target Gene Details
Entrez Gene ID: | 217166 |
Gene Name: | nuclear receptor subfamily 1, group D, member 1 |
Gene Aliases: |
A530070C09Rik, R75201 |
Location: |
Chr.11:98767932-98775377 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nr1d1 | NM_145434.4 | 8 | 2682 | NP_663409.2 |
| AK135640.1 | 8 | 2544 | ||
| AK137398.1 | BAE23342.1 | |||
| AK137582.1 | 8 | 2547 | BAE23418.1 | |
| AK146430.1 | 8 | 2639 | BAE27164.1 | |
| BC008989.1 | 8 | 2127 | AAH08989.1 | |
| CD776678.1 | 1 | 101 |
More Information
Set Membership: |
|

Genomic Map