Assay Details
Target Gene Details
Entrez Gene ID: | 320207 |
Gene Name: | phosphoinositide-3-kinase, regulatory subunit 5, p101 |
Gene Aliases: |
AV230647, F730038I15Rik |
Location: |
Chr.11:68432125-68497849 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pik3r5 | NM_177320.2 | 19 | 3784 | NP_796294.2 |
| AK149847.1 | 19 | 3752 | BAE29119.1 | |
| AK154643.1 | 19 | 3763 | ||
| AK161050.1 | 10 | 2692 | BAE36171.1 |
More Information
Set Membership: |
|

Genomic Map