Assay Details
Target Gene Details
Entrez Gene ID: | 67390 |
Gene Name: | RNA methyltransferase like 1 |
Gene Aliases: |
4833420N02Rik, AI255153, HC90 |
Location: |
Chr.11:76243736-76250625 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnmtl1 | NM_183263.5 | 1 | 231 | NP_899086.2 |
| AK012200.1 | 1 | 207 | ||
| AK014739.1 | 1 | 198 | BAC25446.1 | |
| AK041163.1 | 1 | 215 | ||
| AK132643.1 | 1 | 232 | ||
| AK147805.1 | 1 | 212 | BAE28150.1 | |
| BC038288.1 | 1 | 180 | AAH38288.1 | |
| BC090975.1 | 1 | 203 |
More Information
Set Membership: |
|

Genomic Map