Assay Details
Target Gene Details
Entrez Gene ID: | 67390 |
Gene Name: | RNA methyltransferase like 1 |
Gene Aliases: |
4833420N02Rik, AI255153, HC90 |
Location: |
Chr.11:76243736-76250625 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnmtl1 | NM_183263.5 | 4 | 1369 | NP_899086.2 |
| AK012200.1 | 4 | 1344 | ||
| AK014739.1 | 4 | 1335 | BAC25446.1 | |
| AK147805.1 | 4 | 1350 | BAE28150.1 | |
| BC038288.1 | 4 | 1318 | AAH38288.1 | |
| BC090975.1 | 4 | 1341 | ||
| BC115622.1 | ||||
| BC115623.1 |
More Information
Set Membership: |
|

Genomic Map