Assay Details
Target Gene Details
Entrez Gene ID: | 237987 |
Gene Name: | otopetrin 2 |
Gene Aliases: |
4732464P15 |
Location: |
Chr.11:115307163-115332306 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Otop2 | NM_172801.2 | NP_766389.2 | ||
| AK028866.1 | BAC26159.1 |
Target Gene Details
Entrez Gene ID: | 16470 |
Gene Name: | Usher syndrome 1G |
Gene Aliases: |
Sans, js |
Location: |
Chr.11:115315192-115322142 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ush1g | NM_176847.3 | 2 | 1037 | NP_789817.1 |
| AB087502.1 | 2 | 1037 | BAC57430.1 | |
| BC120509.1 | 2 | 1020 | ||
| BC137807.1 | 2 | 1020 |
More Information
Set Membership: |
|

Genomic Map