Assay Details
Target Gene Details
Entrez Gene ID: | 194908 |
Gene Name: | phospholipase D family, member 6 |
Gene Aliases: |
4933433K01Rik, Gm10, Zuc, mZuc, mitoPLD |
Location: |
Chr.11:59783893-59787657 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pld6 | NM_001290283.1 | 1 | 364 | NP_001277212.1 |
| NM_183139.2 | 1 | 364 | NP_898962.2 | |
| AK077214.1 | 1 | 326 | BAC36687.1 | |
| AK139586.1 | 1 | 352 | BAE24077.1 | |
| BC119245.1 | 1 | 364 | ||
| BC145052.1 | 1 | 364 |
More Information
Set Membership: |
|

Genomic Map