Assay Details
Target Gene Details
Entrez Gene ID: | 68275 |
Gene Name: | replication protein A1 |
Gene Aliases: |
5031405K23Rik, 70kDa, AA589576, AW557552, RF-A, RP-A, Rpa |
Location: |
Chr.11:75300259-75348383 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rpa1 | NM_001164223.1 | 18 | 2673 | NP_001157695.1 |
| NM_026653.2 | 17 | 2610 | NP_080929.1 | |
| AK014298.1 | 6 | 1313 | ||
| AK019864.1 | 11 | 2011 | ||
| AK035084.1 | 17 | 2538 | ||
| AK148035.1 | 17 | 2551 | BAE28302.1 | |
| AK150785.1 | 17 | 2507 | BAE29849.1 | |
| AK151590.1 | 17 | 2563 | BAE30530.1 | |
| AK152296.1 | 18 | 2603 | BAE31105.1 | |
| AK153107.1 | 18 | 2603 | ||
| AK153144.1 | 17 | 2563 | BAE31755.1 | |
| AK165316.1 | 17 | 2523 | BAE38134.1 | |
| AK165944.1 | 17 | 2541 | BAE38476.1 | |
| AK167598.1 | 17 | 2496 | BAE39655.1 | |
| AK168400.1 | 17 | 2563 | ||
| AK169599.1 | 17 | 2534 | BAE41250.1 | |
| AK196893.1 | ||||
| AK202801.1 | ||||
| AK206421.1 | ||||
| AK206963.1 | ||||
| AK206973.1 | ||||
| AK214358.1 | ||||
| AK214387.1 | ||||
| AK216001.1 | ||||
| AK216329.1 | ||||
| AK216964.1 | 1 | 371 | ||
| AK217322.1 | 1 | 371 | ||
| AK218266.1 | ||||
| AK218731.1 | 1 | 371 | ||
| BC019119.1 | 17 | 2541 | AAH19119.1 | |
| BE981198.1 | 1 | 449 |
More Information
Set Membership: |
|

Genomic Map