Assay Details
Target Gene Details
Entrez Gene ID: | 140486 |
Gene Name: | insulin-like growth factor 2 mRNA binding protein 1 |
Gene Aliases: |
AL024068, AW549074, CRD-BP, Crdbp, D030026A21Rik, D11Moh40e, D11Moh45, IMP-1, Neilsen, ZBP-1, Zbp1, mir-3063 |
Location: |
Chr.11:95957163-96005944 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igf2bp1 | NM_009951.4 | 6 | 760 | NP_034081.1 |
| AF061569.1 | 6 | 579 | AAC72743.1 | |
| AK013940.1 | 6 | 727 | BAB29071.1 | |
| AK044850.1 | 6 | 665 | BAC32119.1 | |
| BC051679.1 | 6 | 743 | AAH51679.1 |
More Information
Set Membership: |
|

Genomic Map