Assay Details
Target Gene Details
Entrez Gene ID: | 71833 |
Gene Name: | DDB1 and CUL4 associated factor 7 |
Gene Aliases: |
1700012F10Rik, 2610037L01Rik, C86529, HAN11, Wdr68 |
Location: |
Chr.11:106036872-106059324 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dcaf7 | NM_027946.3 | 1 | 67 | NP_082222.1 |
| AK005902.1 | 1 | 61 | BAB24308.1 | |
| AK047435.1 | 1 | 68 | BAC33058.1 | |
| AK080823.1 | ||||
| AK137813.1 | ||||
| AK154757.1 | 1 | 58 | BAE32807.1 | |
| AK165674.1 | 1 | 58 | BAE38331.1 | |
| BB646282.1 | 1 | 68 | ||
| BC048165.1 | AAH48165.1 | |||
| BC048722.1 | AAH48722.1 |

Genomic Map