Assay Details
Target Gene Details
Entrez Gene ID: | 71833 |
Gene Name: | DDB1 and CUL4 associated factor 7 |
Gene Aliases: |
1700012F10Rik, 2610037L01Rik, C86529, HAN11, Wdr68 |
Location: |
Chr.11:106036872-106059324 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dcaf7 | NM_027946.3 | 7 | 2645 | NP_082222.1 |
| AK154757.1 | 7 | 2636 | BAE32807.1 | |
| AK193740.1 | ||||
| BC048165.1 | 7 | 2558 | AAH48165.1 |

Genomic Map