Assay Details
Target Gene Details
Entrez Gene ID: | 22780 |
Gene Name: | IKAROS family zinc finger 3 |
Gene Aliases: |
5830411O07Rik, Aiolos, Zfpn1a3, Znfn1a3 |
Location: |
Chr.11:98464610-98546380 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ikzf3 | NM_011771.1 | 8 | 2605 | NP_035901.1 |
| AK037261.1 | 1 | 1014 | BAE20507.1 | |
| AK038043.1 | 2 | 3068 | ||
| AK088470.1 | 1 | 315 |
More Information
Set Membership: |
|

Genomic Map