Assay Details
Target Gene Details
Entrez Gene ID: | 320472 |
Gene Name: | protein phosphatase 1E (PP2C domain containing) |
Gene Aliases: |
AW049266, B930008A12Rik, POPX1, PP2CH, mKIAA1072 |
Location: |
Chr.11:87226906-87358994 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppm1e | NM_177167.4 | 1 | 91 | NP_796141.2 |
| AK046962.1 | 1 | 146 | BAC32927.1 | |
| AK053696.1 | BAC35479.1 | |||
| BB665375.1 | ||||
| BC138900.1 | 1 | 58 | ||
| BC138903.1 | 1 | 58 | ||
| BY004447.1 | 1 | 92 |
More Information
Set Membership: |
|

Genomic Map