Assay Details
Target Gene Details
Entrez Gene ID: | 19049 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 1B |
Gene Aliases: |
AU040756, DARPP-32, Darpp32 |
Location: |
Chr.11:98348738-98357795 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r1b | NM_144828.1 | 1 | 363 | NP_659077.1 |
| AB221544.1 | ||||
| BC011122.1 | 1 | 273 | AAH11122.1 | |
| BC026568.1 | 1 | 363 | AAH26568.1 | |
| BC031129.1 | 1 | 111 | AAH31129.1 |
More Information
Set Membership: |
|

Genomic Map