Assay Details
Target Gene Details
Entrez Gene ID: | 52513 |
Gene Name: | DEAD (Asp-Glu-Ala-Asp) box polypeptide 56 |
Gene Aliases: |
2600001H07Rik, D11Ertd619e, Noh61 |
Location: |
Chr.11:6257545-6267729 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ddx56 | NM_026538.3 | 14 | 2155 | NP_080814.1 |
| AK011136.1 | 14 | 2165 | BAB27426.1 | |
| AK076975.1 | 14 | 2155 | BAC36543.1 | |
| AK077953.1 | 9 | 1402 |

Genomic Map