Assay Details
Target Gene Details
Entrez Gene ID: | 170835 |
Gene Name: | inositol polyphosphate 5-phosphatase J |
Gene Aliases: |
Pib5pa, Pipp |
Location: |
Chr.11:3494272-3505567 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Inpp5j | NM_172439.3 | 2 | 761 | NP_766027.2 |
| AK034272.1 | 2 | 762 | BAC28654.1 | |
| AK136908.1 | 2 | 746 | ||
| AK165718.1 | 2 | 1132 | ||
| AK170114.1 | 2 | 727 | BAE41572.1 | |
| AK171029.1 | 2 | 722 | BAE42196.1 | |
| BC022915.1 | 2 | 714 | ||
| BC131634.1 | 2 | 756 | ||
| BC131635.1 | 2 | 753 |
More Information
Set Membership: |
|

Genomic Map