Assay Details
Target Gene Details
Entrez Gene ID: | 171210 |
Gene Name: | acyl-CoA thioesterase 2 |
Gene Aliases: |
AA571646, MTE-I, Mte1 |
Location: |
Chr.12:83987861-83993877 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acot2 | NM_134188.3 | 1 | 101 | NP_598949.3 |
| AK141651.1 | 1 | 91 | ||
| AK172617.1 | 1 | 97 | BAE43095.1 | |
| BC064469.1 | 1 | 72 | AAH64469.1 |
More Information
Set Membership: |
|

Genomic Map