Assay Details
Target Gene Details
Entrez Gene ID: | 171282 |
Gene Name: | acyl-CoA thioesterase 4 |
Gene Aliases: |
B430212I04Rik, PTE-Ib, Pte1b, Pte2b |
Location: |
Chr.12:84038379-84044723 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acot4 | NM_134247.3 | 3 | 1561 | NP_599008.3 |
| AK046630.1 | 3 | 1561 | BAC32814.1 | |
| AK050420.1 | 3 | 1543 | BAC34246.1 | |
| AK080883.1 | 3 | 1547 | BAC38060.1 | |
| BC113185.1 | 3 | 1505 |
More Information
Set Membership: |
|

Genomic Map