Assay Details
Target Gene Details
Entrez Gene ID: | 12775 |
Gene Name: | chemokine (C-C motif) receptor 7 |
Gene Aliases: |
CC-CKR-7, CCR-7, CD197, Cdw197, Cmkbr7, EBI1, Ebi1h |
Location: |
Chr.11:99144199-99155077 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccr7 | NM_001301713.1 | 2 | 389 | NP_001288642.1 |
| NM_007719.2 | 3 | 439 | NP_031745.2 | |
| AK037965.1 | 3 | 440 | BAC29909.1 | |
| AK154349.1 | 3 | 440 | BAE32532.1 | |
| AK154736.1 | 3 | 440 | BAE32796.1 | |
| AK154849.1 | 3 | 440 | BAE32875.1 |

Genomic Map