Assay Details
Target Gene Details
Entrez Gene ID: | 12464 |
Gene Name: | chaperonin containing Tcp1, subunit 4 (delta) |
Gene Aliases: |
2610204B21Rik, A45, C78323, Cctd |
Location: |
Chr.11:22990593-23003336 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cct4 | NM_009837.1 | 1 | 100 | NP_033967.1 |
| AK010634.1 | 1 | 68 | BAB27078.1 | |
| AK146242.1 | 1 | 70 | BAE27006.1 | |
| AK150601.1 | BAE29693.1 | |||
| AK152173.1 | BAE31005.1 | |||
| AK152665.1 | 1 | 68 | BAE31400.1 | |
| AK163369.1 | ||||
| AK165229.1 | 1 | 68 | BAE38089.1 | |
| AK167026.1 | 1 | 68 | BAE39199.1 | |
| AK167262.1 | BAE39376.1 | |||
| AK167657.1 | ||||
| AK167846.1 | BAE39866.1 | |||
| AK169339.1 | BAE41091.1 | |||
| BC054773.1 | AAH54773.1 | |||
| Z31554.1 | 1 | 100 | CAA83429.1 |
More Information
Set Membership: |
|

Genomic Map